Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024379.hg.1 |
TC05001788.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024379.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024379.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
ENST00000395009.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
ENST00000395009.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012070.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017530.hg.1 |
ENST00000460287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017530.hg.1 |
ENST00000460287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017530.hg.1 |
ENST00000460287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017530.hg.1 |
ENST00000460287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183558.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183558.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183558.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183558.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183558.hg.1 |
TC0500012070.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
ENST00000395009.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
ENST00000395009.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
ENST00000458198.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
NM_080656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012070.hg.1 |
NM_080656.2 |
Refseq |