Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058522.hg.1 |
TC01003832.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058522.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058522.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058522.hg.1 |
ENST00000536446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058522.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231338.hg.1 |
TC0100017358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100231341.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231341.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231341.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231341.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231341.hg.1 |
TC0100017358.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000356609.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000366926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
ENST00000484239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NM_018713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017358.hg.1 |
NR_046437.1 |
Refseq |