Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010897.hg.1 |
TC02000705.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010897.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010897.hg.1 |
ENST00000409871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010897.hg.1 |
ENST00000542974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010897.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010892.hg.1 |
TC02000705.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010892.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010892.hg.1 |
ENST00000409871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010892.hg.1 |
ENST00000542974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010892.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010895.hg.1 |
TC02000705.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010895.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010895.hg.1 |
ENST00000409871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010895.hg.1 |
ENST00000542974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010895.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010894.hg.1 |
TC02000705.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010894.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010894.hg.1 |
ENST00000409871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010894.hg.1 |
ENST00000542974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010894.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010890.hg.1 |
TC02000705.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010890.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010890.hg.1 |
ENST00000409871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010890.hg.1 |
ENST00000542974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010890.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000607612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
ENST00000607612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162210.hg.1 |
TC0200008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200162205.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162205.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162205.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162205.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162205.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162205.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162205.hg.1 |
TC0200008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162209.hg.1 |
TC0200008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000502881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000607612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
ENST00000607612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008911.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
ENST00000476511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162208.hg.1 |
TC0200008911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200162204.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162204.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162204.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162204.hg.1 |
ENST00000343936.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162204.hg.1 |
ENST00000409871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162204.hg.1 |
NM_198581.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162204.hg.1 |
TC0200008911.hg.1 |
Clariom D TC |