Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021704.hg.1 |
TC09001691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021704.hg.1 |
ENST00000371897.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021704.hg.1 |
ENST00000371899.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021704.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021704.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021704.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011850.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000371897.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000371899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000414172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000432868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
ENST00000485978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
NM_001145099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
NM_017585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181156.hg.1 |
TC0900011850.hg.1 |
Clariom D TC |