Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027580.hg.1 |
TC03001515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027580.hg.1 |
ENST00000283268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027580.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027580.hg.1 |
ENST00000486811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027580.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027584.hg.1 |
TC03001515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027584.hg.1 |
ENST00000283268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027584.hg.1 |
ENST00000486811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027584.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027585.hg.1 |
TC03001515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027585.hg.1 |
ENST00000283268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027585.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027578.hg.1 |
TC03001515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027578.hg.1 |
ENST00000283268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027578.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027578.hg.1 |
ENST00000486811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027578.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027579.hg.1 |
TC03001515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027579.hg.1 |
ENST00000283268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027579.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027579.hg.1 |
ENST00000486811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027579.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027583.hg.1 |
TC03001515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027583.hg.1 |
ENST00000283268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027583.hg.1 |
ENST00000486811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027583.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011362.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185183.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185183.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185183.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185183.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185183.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185183.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185183.hg.1 |
TC0300011362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185178.hg.1 |
TC0300011362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185182.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185182.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185182.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185182.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185182.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185182.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185182.hg.1 |
TC0300011362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185184.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185184.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185184.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185184.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185184.hg.1 |
TC0300011362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185177.hg.1 |
TC0300011362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185179.hg.1 |
TC0300011362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
ENST00000283268.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
ENST00000475839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
ENST00000486811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
NM_018008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185180.hg.1 |
TC0300011362.hg.1 |
Clariom D TC |