Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011513013.2 | XP_011511315.2 | 9 - 10 | 4827 | 157 | |
XM_017006898.1 | XP_016862387.1 | 10 - 11 | 3066 | 157 | ||
XM_017006901.1 | XP_016862390.1 | 10 - 11 | 4700 | 157 | ||
XM_017006905.1 | XP_016862394.1 | 11 - 12 | 4283 | 157 | ||
XM_017006906.1 | XP_016862395.1 | 11 - 12 | 4064 | 157 | ||
XM_017006907.1 | XP_016862396.1 | 11 - 12 | 3829 | 157 | ||
XM_017006908.1 | XP_016862397.1 | 11 - 12 | 3954 | 157 | ||
XM_017006909.1 | XP_016862398.1 | 10 - 11 | 3741 | 157 | ||
XM_017006912.1 | XP_016862401.1 | 9 - 10 | 3482 | 157 | ||
XM_017006914.1 | XP_016862403.1 | 9 - 10 | 3689 | 157 | ||
XM_017006915.1 | XP_016862404.1 | 9 - 10 | 3671 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |