Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011513013.2 | XP_011511315.2 | 10 - 11 | 5036 | 101 | |
XM_011513019.2 | XP_011511321.2 | 9 - 10 | 2396 | 101 | ||
XM_011513020.2 | XP_011511322.2 | 10 - 11 | 2459 | 101 | ||
XM_017006898.1 | XP_016862387.1 | 11 - 12 | 3275 | 101 | ||
XM_017006899.1 | XP_016862388.1 | 11 - 12 | 5099 | 101 | ||
XM_017006900.1 | XP_016862389.1 | 10 - 11 | 3172 | 101 | ||
XM_017006901.1 | XP_016862390.1 | 11 - 12 | 4909 | 101 | ||
XM_017006902.1 | XP_016862391.1 | 11 - 12 | 3235 | 101 | ||
XM_017006905.1 | XP_016862394.1 | 12 - 13 | 4492 | 101 | ||
XM_017006906.1 | XP_016862395.1 | 12 - 13 | 4273 | 101 | ||
XM_017006907.1 | XP_016862396.1 | 12 - 13 | 4038 | 101 | ||
XM_017006908.1 | XP_016862397.1 | 12 - 13 | 4163 | 101 | ||
XM_017006909.1 | XP_016862398.1 | 11 - 12 | 3950 | 101 | ||
XM_017006912.1 | XP_016862401.1 | 10 - 11 | 3691 | 101 | ||
XM_017006914.1 | XP_016862403.1 | 10 - 11 | 3898 | 101 | ||
XM_017006915.1 | XP_016862404.1 | 10 - 11 | 3880 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |