Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006509.hg.1 |
TC01000424.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006509.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006509.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153912.hg.1 |
TC0100007672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153915.hg.1 |
TC0100007672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153913.hg.1 |
TC0100007672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000355082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000373586.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000471486.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000474371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000483517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
ENST00000489353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007672.hg.1 |
NM_003757.3 |
Refseq |