Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014877.3 | NP_055692.2 | 25 - 26 | 3629 | 139 | |
XM_005257888.4 | XP_005257945.1 | 26 - 27 | 3758 | 139 | ||
XM_005257889.4 | XP_005257946.1 | 25 - 26 | 3680 | 139 | ||
XM_006722214.3 | XP_006722277.1 | 26 - 27 | 3761 | 139 | ||
XM_006722215.3 | XP_006722278.1 | 24 - 25 | 3633 | 139 | ||
XM_006722216.3 | XP_006722279.1 | 15 - 16 | 2463 | 139 | ||
XM_011525544.2 | XP_011523846.1 | 26 - 27 | 3832 | 139 | ||
XM_017025477.1 | XP_016880966.1 | 23 - 24 | 3549 | 139 | ||
XM_017025478.1 | XP_016880967.1 | 26 - 27 | 3814 | 139 | ||
GenBank mRNA | BC094881.1 | - | 23 - 24 | 3655 | 3076113 | 139 |
BC130582.1 | - | 25 - 26 | 3609 | 40146613 | 139 | |
BC144083.1 | - | 25 - 26 | 3612 | 9052603 | 139 | |
D29677.2 | - | 24 - 25 | 3587 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |