Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290263.1 | NP_001277192.1 | 48 - 49 | 5514 | 89 | |
NM_014689.2 | NP_055504.2 | 48 - 49 | 5657 | 89 | ||
XM_005246673.4 | XP_005246730.1 | 48 - 49 | 5516 | 89 | ||
XM_006712617.3 | XP_006712680.1 | 48 - 49 | 5681 | 89 | ||
XM_017004441.1 | XP_016859930.1 | 48 - 49 | 5684 | 89 | ||
XM_017004442.1 | XP_016859931.1 | 48 - 49 | 5660 | 89 | ||
XM_017004443.1 | XP_016859932.1 | 48 - 49 | 5733 | 89 | ||
XM_017004444.1 | XP_016859933.1 | 47 - 48 | 5640 | 89 | ||
XM_017004445.1 | XP_016859934.1 | 48 - 49 | 5514 | 89 | ||
XM_017004446.1 | XP_016859935.1 | 47 - 48 | 5538 | 89 | ||
XM_017004447.1 | XP_016859936.1 | 48 - 49 | 5656 | 89 | ||
GenBank mRNA | AB074179.1 | - | 3 - 4 | 247 | 89 | |
AF250396.1 | - | 7 - 8 | 927 | 89 | ||
AK000227.1 | - | 6 - 7 | 626 | 89 | ||
AK001253.1 | - | 3 - 4 | 257 | 89 | ||
AK293847.1 | - | 14 - 15 | 1583 | 89 | ||
AK293879.1 | - | 18 - 19 | 1964 | 89 | ||
AK307914.1 | - | 8 - 9 | 1018 | 89 | ||
BC015018.1 | - | 6 - 7 | 624 | 89 | ||
CR749492.1 | - | 23 - 24 | 2502 | 89 | ||
EU236710.2 | - | 48 - 49 | 5511 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |