Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017007235.1 | XP_016862724.1 | 2 - 3 | 245 | 84 | |
XM_017007236.1 | XP_016862725.1 | 2 - 3 | 233 | 84 | ||
GenBank mRNA | AK023029.1 | - | 2 - 3 | 209 | 84 | |
BC070164.1 | - | 2 - 3 | 213 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |