Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037177.hg.1 |
TC01002409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037177.hg.1 |
ENST00000247087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037177.hg.1 |
ENST00000374011.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037177.hg.1 |
ENST00000482400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037177.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000480033.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000480033.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000480033.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037178.hg.1 |
TC01002409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037178.hg.1 |
ENST00000247087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037178.hg.1 |
ENST00000374011.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037178.hg.1 |
ENST00000482400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037178.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037176.hg.1 |
TC01002409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037176.hg.1 |
ENST00000374011.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037176.hg.1 |
ENST00000482400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037176.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000247087.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000480033.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000480033.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000480033.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000487743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
ENST00000490295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013437.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199844.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199844.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199844.hg.1 |
ENST00000374011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199844.hg.1 |
NM_001029882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199844.hg.1 |
TC0100013437.hg.1 |
Clariom D TC |