Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022113.hg.1 |
TC05001620.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022113.hg.1 |
ENST00000274382.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022113.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179911.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179911.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179911.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179911.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179911.hg.1 |
TC0500011528.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000274382.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
ENST00000512378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011528.hg.1 |
NM_153234.4 |
Refseq |