Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031648.hg.1 |
TC03001741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031648.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031648.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031650.hg.1 |
TC03001741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031650.hg.1 |
ENST00000290868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031650.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031650.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031650.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191912.hg.1 |
TC0300012282.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000290868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012282.hg.1 |
NM_144639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191910.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191910.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191910.hg.1 |
ENST00000383579.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191910.hg.1 |
NM_001165974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191910.hg.1 |
TC0300012282.hg.1 |
Clariom D TC |