Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018709.hg.1 |
TC03001071.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018709.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018709.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300172091.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300172091.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300172091.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300172091.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300172091.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300172091.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300172091.hg.1 |
TC0300009916.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000232424.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
ENST00000476918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009916.hg.1 |
NM_005524.3 |
Refseq |