Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021552.hg.1 |
TC06001785.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021552.hg.1 |
ENST00000263045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021552.hg.1 |
ENST00000371159.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021552.hg.1 |
ENST00000393666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021552.hg.1 |
ENST00000433368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021552.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021552.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021566.hg.1 |
TC06001785.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021566.hg.1 |
ENST00000263045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021566.hg.1 |
ENST00000371159.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021566.hg.1 |
ENST00000423399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021566.hg.1 |
ENST00000433368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021566.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021566.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021551.hg.1 |
TC06001785.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021551.hg.1 |
ENST00000263045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021551.hg.1 |
ENST00000371159.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021551.hg.1 |
ENST00000393666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021551.hg.1 |
ENST00000433368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021551.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021551.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021565.hg.1 |
TC06001785.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021565.hg.1 |
ENST00000263045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021565.hg.1 |
ENST00000371159.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021565.hg.1 |
ENST00000423399.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021565.hg.1 |
ENST00000433368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021565.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021565.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021567.hg.1 |
TC06001785.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021567.hg.1 |
ENST00000263045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021567.hg.1 |
ENST00000371159.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021567.hg.1 |
ENST00000433368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021567.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021567.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021562.hg.1 |
TC06001785.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021562.hg.1 |
ENST00000263045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021562.hg.1 |
ENST00000433368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021562.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021562.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182592.hg.1 |
TC0600012003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182593.hg.1 |
TC0600012003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182591.hg.1 |
TC0600012003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182599.hg.1 |
TC0600012003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000263045.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000354620.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000371159.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
ENST00000433368.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_001190986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012003.hg.1 |
NM_006061.3 |
Refseq |