Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB067492.1 | - | 11 - 12 | 1788 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |