Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000265577.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000317367.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000339809.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000396675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000401396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000402384.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000406470.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000407906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000422063.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000430867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000446305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000447326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000455539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000457755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_001136020.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_001276478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_004968.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_022307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000265577.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000317367.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000339809.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000396675.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000401396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000402384.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000406470.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000407906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000422063.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000430867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000446305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000447326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000455539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
ENST00000457755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_001136020.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_001276478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_004968.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
NM_022307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173006.hg.1 |
TC0700010257.hg.1 |
Clariom D TC |