Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024129.hg.1 |
ENST00000221978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024129.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024129.hg.1 |
TC19001776.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024131.hg.1 |
ENST00000221978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024131.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024131.hg.1 |
TC19001776.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024130.hg.1 |
ENST00000221978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024130.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024130.hg.1 |
TC19001776.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
TC1900011251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194701.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194704.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194704.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194704.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194704.hg.1 |
TC1900011251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194704.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194704.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194704.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194698.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194698.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194698.hg.1 |
TC1900011251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194698.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194698.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
TC1900011251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194699.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
TC1900011251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194702.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000221978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000593572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000595217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
ENST00000600427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011251.hg.1 |
NM_005601.3 |
Refseq |