Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Hs01125363_m1
See other NRXN2 GE Assays ›
Gene Symbol:
NRXN2
Gene Name:
neurexin 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 64606174 - 64723188 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01125363_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: NRXN2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_015080.3
NP_055895.1
17 - 18
3869
77
NM_138732.2
NP_620060.1
15 - 16
3749
77
XM_005274400.3
XP_005274457.1
16 - 17
3401
77
XM_005274401.3
XP_005274458.1
16 - 17
3401
77
XM_005274402.3
XP_005274459.1
15 - 16
3377
77
XM_011545370.1
XP_011543672.1
16 - 17
3431
77
XM_011545371.1
XP_011543673.1
16 - 17
3428
77
XM_011545373.1
XP_011543675.1
16 - 17
3410
77
XM_011545375.1
XP_011543677.1
15 - 16
3386
77
XM_011545385.1
XP_011543687.1
9 - 10
1776
77
XM_017018563.1
XP_016874052.1
15 - 16
3401
77
XM_017018564.1
XP_016874053.1
15 - 16
3398
77
XM_017018565.1
XP_016874054.1
15 - 16
3395
77
XM_017018566.1
XP_016874055.1
15 - 16
3359
77
XM_017018567.1
XP_016874056.1
16 - 17
3431
77
XM_017018568.1
XP_016874057.1
16 - 17
3422
77
XM_017018569.1
XP_016874058.1
14 - 15
3329
77
XM_017018570.1
XP_016874059.1
16 - 17
3431
77
XM_017018571.1
XP_016874060.1
16 - 17
3422
77
XM_017018572.1
XP_016874061.1
16 - 17
2726
77
XM_017018573.1
XP_016874062.1
15 - 16
3395
77
XM_017018574.1
XP_016874063.1
15 - 16
2696
77
XM_017018575.1
XP_016874064.1
15 - 16
2693
77
XM_017018576.1
XP_016874065.1
11 - 12
2341
77
XM_017018577.1
XP_016874066.1
9 - 10
1767
77
XM_017018578.1
XP_016874067.1
16 - 17
3422
77
GenBank mRNA
AB023138.2
-
16 - 17
3726
77
AB035266.1
-
16 - 17
3521
77
AB177862.1
-
13 - 14
2573
77
BC150275.1
-
16 - 17
3726
9007249
77
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene NRXN2
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
signal transduction
chemical synaptic transmission
neurotransmitter secretion
synapse assembly
adult behavior
social behavior
vocal learning
vocalization behavior
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
transmembrane signaling receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0