Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007118.3 | NP_009049.2 | 48 - 49 | 7661 | 130 | |
XM_011514107.2 | XP_011512409.1 | 48 - 49 | 8236 | 130 | ||
XM_011514108.1 | XP_011512410.1 | 48 - 49 | 7535 | 130 | ||
XM_011514109.2 | XP_011512411.1 | 48 - 49 | 11313 | 130 | ||
XM_011514110.2 | XP_011512412.1 | 48 - 49 | 7771 | 130 | ||
XM_017009801.1 | XP_016865290.1 | 48 - 49 | 8013 | 130 | ||
XM_017009802.1 | XP_016865291.1 | 48 - 49 | 8013 | 130 | ||
XM_017009803.1 | XP_016865292.1 | 41 - 42 | 6282 | 130 | ||
GenBank mRNA | AF091395.1 | - | 48 - 49 | 7526 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |