Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000623525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014302.hg.1 |
ENST00000606125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000623525.1 |
Ensembl |