Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167856.1 | NP_001161328.1 | 27 - 28 | 3625 | 127 | |
NM_018183.3 | NP_060653.3 | 27 - 28 | 3622 | 127 | ||
XM_005253572.3 | XP_005253629.1 | 28 - 29 | 3763 | 127 | ||
XM_005253573.4 | XP_005253630.1 | 29 - 30 | 4173 | 127 | ||
XM_005253575.3 | XP_005253632.1 | 28 - 29 | 3760 | 127 | ||
XM_005253576.3 | XP_005253633.1 | 29 - 30 | 3804 | 127 | ||
XM_006719473.3 | XP_006719536.1 | 30 - 31 | 3929 | 127 | ||
XM_006719474.3 | XP_006719537.1 | 29 - 30 | 3882 | 127 | ||
XM_011538533.2 | XP_011536835.1 | 30 - 31 | 3923 | 127 | ||
XM_011538534.2 | XP_011536836.1 | 27 - 28 | 3661 | 127 | ||
XM_011538535.2 | XP_011536837.1 | 27 - 28 | 3663 | 127 | ||
XM_017019556.1 | XP_016875045.1 | 28 - 29 | 3747 | 127 | ||
XM_017019557.1 | XP_016875046.1 | 28 - 29 | 3754 | 127 | ||
XM_017019558.1 | XP_016875047.1 | 29 - 30 | 3817 | 127 | ||
XM_017019559.1 | XP_016875048.1 | 28 - 29 | 3884 | 127 | ||
GenBank mRNA | AB014772.1 | - | 27 - 28 | 3622 | 127 | |
AK001563.1 | - | 4 - 5 | 368 | 127 | ||
AK096864.1 | - | 10 - 11 | 995 | 127 | ||
BC133704.1 | - | 27 - 28 | 3625 | 40124902 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |