Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000369893.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000459894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000480358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602439.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
ENST00000602868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008749.hg.1 |
NM_178858.5 |
Refseq |