Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007323.hg.1 |
ENST00000216211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007323.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007323.hg.1 |
TC22000379.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
ENST00000396082.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_001167574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007612.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154031.hg.1 |
TC2200007612.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154031.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154031.hg.1 |
NM_006953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154031.hg.1 |
ENST00000216211.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154031.hg.1 |
NM_006953.3 |
Refseq |