Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022435.hg.1 |
TC09001743.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022435.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022435.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022434.hg.1 |
TC09001743.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022434.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022434.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182415.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182415.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182415.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182415.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182415.hg.1 |
TC0900012006.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012006.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182413.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182413.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182413.hg.1 |
ENST00000414656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182413.hg.1 |
NR_024580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182413.hg.1 |
TC0900012006.hg.1 |
Clariom D TC |