Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000451479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000451479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
ENST00000451479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000343605.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000406566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000451479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000451479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
ENST00000451479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007068.hg.1 |
NM_001001479.3 |
Refseq |