Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231381 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952320 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952318 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231497 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231384 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952219 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231490 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952272 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952508 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952497 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952506 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231485 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952266 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952270 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231489 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16067665 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231498 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952510 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952520 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231477 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952512 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231443 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231431 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952295 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952276 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231486 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952500 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231422 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952280 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952334 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16067642 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952513 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952296 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231500 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231457 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952496 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231696 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952285 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231505 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952327 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952511 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231481 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952317 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952265 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952515 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952224 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952260 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952291 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952519 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952275 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231482 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231684 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231447 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231385 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952299 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952290 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952271 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231458 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231504 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952509 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952333 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231434 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952326 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231484 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231679 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952499 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952316 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952223 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231424 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231427 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231677 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952278 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952339 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952283 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231483 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952336 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952287 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952268 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952286 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16067623 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952298 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952263 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231686 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231503 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231464 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231440 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952261 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952226 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952311 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952505 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952504 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05952312 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26231507 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
175365 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
19316 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
15510 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
175041 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
45291 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
30316 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
752974 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
581192 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
246959 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
790291 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
962036 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
493341 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
270787 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
341501 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
424160 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
468819 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
751881 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
928903 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
871864 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
222364 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
894832 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
871700 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
133770 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
384934 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
319892 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
238411 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
86776 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
152621 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
285924 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3373014 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
40153 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P38303563 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01861086 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01861052 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P59749061 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P18213978 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P16380649 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3373014 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178391 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
116860 |
BC008818.2 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
116860 |
CR602359.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
116860 |
NM_003377.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
116860 |
U43368.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
116860 |
U43369.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
116860 |
U48801.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
116860 |
U52819.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
26366 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18213981 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861126 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861052 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P38303563 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861073 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18213858 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18213865 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18213890 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861117 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861066 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18213870 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18213866 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P38303581 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P25919357 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861092 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861086 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01861091 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18213978 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P16381425 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P18213981 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P01861052 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P18213978 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P18213890 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_18_P16381425 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P01861086 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P01861066 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P18213858 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P18213866 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P38303563 |
- |
- |