Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015883.hg.1 |
TC0X001178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015883.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015883.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015884.hg.1 |
TC0X001178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015884.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015884.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015886.hg.1 |
TC0X001178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015886.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015886.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169252.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169252.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169252.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169252.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169252.hg.1 |
TC0X00010144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010144.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169251.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169251.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169251.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169251.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169251.hg.1 |
TC0X00010144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00169254.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169254.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169254.hg.1 |
ENST00000321110.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169254.hg.1 |
NM_152694.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169254.hg.1 |
TC0X00010144.hg.1 |
Clariom D TC |