Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040922.hg.1 |
TC01002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040922.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040922.hg.1 |
ENST00000481882.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040922.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040913.hg.1 |
TC01002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040913.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040913.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040915.hg.1 |
TC01002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040915.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040915.hg.1 |
ENST00000481882.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040915.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040915.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040917.hg.1 |
TC01002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040917.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040917.hg.1 |
ENST00000481882.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040917.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040917.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040912.hg.1 |
TC01002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040912.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040916.hg.1 |
TC01002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040916.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040916.hg.1 |
ENST00000481882.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040916.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040916.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040914.hg.1 |
TC01002621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040914.hg.1 |
ENST00000481882.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040914.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01040914.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205139.hg.1 |
TC0100014058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205145.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205145.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205145.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205145.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205145.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205145.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205145.hg.1 |
TC0100014058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205141.hg.1 |
TC0100014058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205138.hg.1 |
TC0100014058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014058.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205136.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205136.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205136.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205136.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205136.hg.1 |
TC0100014058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000396314.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000481882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205140.hg.1 |
TC0100014058.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205137.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205137.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205137.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205137.hg.1 |
ENST00000294445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205137.hg.1 |
ENST00000524908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205137.hg.1 |
NM_001097611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205137.hg.1 |
TC0100014058.hg.1 |
Clariom D TC |