Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013516.hg.1 |
TC14002326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013516.hg.1 |
ENST00000281581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013516.hg.1 |
ENST00000537690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013516.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182614.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182614.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182614.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182614.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182614.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182614.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182614.hg.1 |
TC1400010758.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000281581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000537690.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000553317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
ENST00000556996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010758.hg.1 |
NM_001164399.1 |
Refseq |