Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
ENST00000342528.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
ENST00000399391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
ENST00000399397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
NM_001277269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
NM_001292063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
ENST00000342528.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
ENST00000399391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
ENST00000399397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
NM_001277269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
NM_001292063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149088.hg.1 |
TC1100006973.hg.1 |
Clariom D TC |