Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000335247.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
ENST00000409327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014164.hg.1 |
NM_001001665.3 |
Refseq |