Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000327.hg.1 |
TC05000016.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000327.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000327.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144668.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144668.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144668.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144668.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144668.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144668.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144668.hg.1 |
TC0500006493.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000304460.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
ENST00000515652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006493.hg.1 |
NM_001003841.2 |
Refseq |