Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017250.hg.1 |
ENST00000334574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017250.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017250.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017250.hg.1 |
TC15001759.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000334574.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000541889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
ENST00000561368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001007122.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010249.hg.1 |
NM_001281806.1 |
Refseq |