Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001540.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001540.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001540.hg.1 |
TC12000110.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001541.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001541.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001541.hg.1 |
TC12000110.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200146751.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146751.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146751.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146751.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146751.hg.1 |
TC1200006676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
ENST00000533292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006676.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146752.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146752.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146752.hg.1 |
ENST00000399422.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146752.hg.1 |
NM_001080454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146752.hg.1 |
TC1200006676.hg.1 |
Clariom D TC |