Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172318.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172318.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172318.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172318.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172318.hg.1 |
TC1000010597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010597.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172316.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172316.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172316.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172316.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172316.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172316.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172316.hg.1 |
TC1000010597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000172315.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172315.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172315.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172315.hg.1 |
ENST00000374139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172315.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172315.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172315.hg.1 |
TC1000010597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000172319.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172319.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172319.hg.1 |
ENST00000434016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172319.hg.1 |
NM_001276451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172319.hg.1 |
TC1000010597.hg.1 |
Clariom D TC |