Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291661.1 | NP_001278590.1 | 30 - 31 | 4826 | 125 | |
XM_011533466.1 | XP_011531768.1 | 30 - 31 | 4823 | 125 | ||
XM_011533467.1 | XP_011531769.1 | 30 - 31 | 4790 | 125 | ||
XM_011533468.1 | XP_011531770.1 | 30 - 31 | 4823 | 125 | ||
XM_011533469.1 | XP_011531771.1 | 30 - 31 | 4823 | 125 | ||
XM_011533471.2 | XP_011531773.1 | 30 - 31 | 4823 | 125 | ||
XM_017005860.1 | XP_016861349.1 | 30 - 31 | 4823 | 125 | ||
XM_017005861.1 | XP_016861350.1 | 29 - 30 | 5506 | 125 | ||
XM_017005862.1 | XP_016861351.1 | 9 - 10 | 1398 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |