Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020759.2 | NP_065810.2 | 29 - 30 | 13614 | 85 | |
XM_011521831.2 | XP_011520133.1 | 29 - 30 | 13627 | 85 | ||
XM_011521832.2 | XP_011520134.1 | 29 - 30 | 13624 | 85 | ||
XM_011521833.2 | XP_011520135.1 | 28 - 29 | 13594 | 85 | ||
XM_011521834.2 | XP_011520136.1 | 28 - 29 | 13564 | 85 | ||
XM_011521835.2 | XP_011520137.1 | 27 - 28 | 13486 | 85 | ||
XM_011521836.2 | XP_011520138.1 | 29 - 30 | 13929 | 85 | ||
XM_011521837.2 | XP_011520139.1 | 27 - 28 | 13396 | 85 | ||
XM_011521839.2 | XP_011520141.1 | 16 - 17 | 12396 | 85 | ||
XM_017022439.1 | XP_016877928.1 | 28 - 29 | 13549 | 85 | ||
XM_017022440.1 | XP_016877929.1 | 28 - 29 | 13526 | 85 | ||
XM_017022441.1 | XP_016877930.1 | 27 - 28 | 13845 | 85 | ||
XM_017022442.1 | XP_016877931.1 | 22 - 23 | 12953 | 85 | ||
XM_017022443.1 | XP_016877932.1 | 21 - 22 | 12937 | 85 | ||
GenBank mRNA | AB037721.1 | - | 7 - 8 | 4918 | 85 | |
AK297603.1 | - | 7 - 8 | 1134 | 85 | ||
AL133579.1 | - | 3 - 4 | 261 | 85 | ||
CR749416.1 | - | 7 - 8 | 7636 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |