Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002336.hg.1 |
ENST00000219905.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002336.hg.1 |
ENST00000389936.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002336.hg.1 |
ENST00000545763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002336.hg.1 |
NM_001080541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002336.hg.1 |
NM_001164273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002336.hg.1 |
TC15000311.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000219905.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000545763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000564190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000566586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000568255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000570161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
NM_001080541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
NM_001164273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000219905.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000545763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000564190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000566586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000568255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
ENST00000570161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
NM_001080541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
NM_001164273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149133.hg.1 |
TC1500007009.hg.1 |
Clariom D TC |