Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021842.hg.1 |
TC11003485.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021842.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021842.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021844.hg.1 |
TC11003485.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021844.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021844.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021843.hg.1 |
TC11003485.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021843.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021843.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204270.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204270.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204270.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204270.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204270.hg.1 |
TC1100013172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100204269.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204269.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204269.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204269.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204269.hg.1 |
TC1100013172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100204271.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204271.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204271.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204271.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204271.hg.1 |
TC1100013172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
ENST00000330574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013172.hg.1 |
NM_030628.1 |
Refseq |