Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC036938.1 | - | 3 - 4 | 598 | 89 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | F-box and leucine rich repeat protein 13 | FBXL13 |
DRC6, Fbl13 |
Anti-sense | Chr. 7 - 102699226 - 103074841 on NCBI Build GRCh38 | |
Downstream | armadillo repeat containing 10 | ARMC10 |
PNAS-112, PNAS112, PSEC0198, SVH |
85.9kb | Sense | Chr. 7 - 103074727 - 103099763 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |