Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020319.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020319.hg.1 |
ENST00000535447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020319.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020319.hg.1 |
TC10001580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020321.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020321.hg.1 |
TC10001580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020318.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020318.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020318.hg.1 |
ENST00000535447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020318.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020318.hg.1 |
TC10001580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020315.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020315.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020315.hg.1 |
ENST00000543866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020315.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020315.hg.1 |
TC10001580.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000178986.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178986.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178986.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178986.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178986.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178986.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178986.hg.1 |
TC1000011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000178988.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178988.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178988.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178988.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178988.hg.1 |
TC1000011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178989.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178989.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178989.hg.1 |
TC1000011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000178985.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178985.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178985.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178985.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178985.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178985.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178985.hg.1 |
TC1000011585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000178987.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178987.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178987.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178987.hg.1 |
ENST00000370508.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178987.hg.1 |
ENST00000471741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178987.hg.1 |
NM_002079.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178987.hg.1 |
TC1000011585.hg.1 |
Clariom D TC |