Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000365427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000462465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000462465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000462465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007349.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007349.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007349.hg.1 |
TC10000657.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007350.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007350.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007350.hg.1 |
TC10000657.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007353.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007353.hg.1 |
ENST00000467521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007353.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007353.hg.1 |
TC10000657.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000157473.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157473.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157473.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157473.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157473.hg.1 |
TC1000008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000365427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000462465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000462465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000462465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
ENST00000492319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008467.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157475.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157475.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157475.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157475.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157475.hg.1 |
TC1000008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000157474.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157474.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157474.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157474.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157474.hg.1 |
TC1000008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000157477.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157477.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157477.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157477.hg.1 |
ENST00000358935.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157477.hg.1 |
ENST00000467521.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157477.hg.1 |
NM_017824.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157477.hg.1 |
TC1000008467.hg.1 |
Clariom D TC |