Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022451.hg.1 |
ENST00000286574.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022451.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022451.hg.1 |
TC12001681.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022452.hg.1 |
ENST00000286574.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022452.hg.1 |
ENST00000535025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022452.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022452.hg.1 |
TC12001681.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200186279.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186279.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186279.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186279.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186279.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186279.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186279.hg.1 |
TC1200011108.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |