Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003826.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003826.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003826.hg.1 |
TC16000259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003827.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003827.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16003827.hg.1 |
TC16000259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152085.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152085.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152085.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152085.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152085.hg.1 |
TC1600007238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600152087.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152087.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152087.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152087.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152087.hg.1 |
TC1600007238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007238.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152086.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152086.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152086.hg.1 |
ENST00000300113.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152086.hg.1 |
NM_022097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152086.hg.1 |
TC1600007238.hg.1 |
Clariom D TC |