Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004772.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004772.hg.1 |
ENST00000395086.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004772.hg.1 |
ENST00000395087.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004772.hg.1 |
ENST00000486754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004772.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004772.hg.1 |
TC10000418.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004771.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004771.hg.1 |
ENST00000395086.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004771.hg.1 |
ENST00000395087.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004771.hg.1 |
ENST00000486754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004771.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004771.hg.1 |
TC10000418.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000470920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000470920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000470920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153451.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153451.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153451.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153451.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153451.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153451.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153451.hg.1 |
TC1000007890.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000153452.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153452.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153452.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153452.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153452.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153452.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153452.hg.1 |
TC1000007890.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000354624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000470920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000470920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000470920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
ENST00000488706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007890.hg.1 |
NM_025130.3 |
Refseq |