Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006983.hg.1 |
TC0X000521.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006983.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006983.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006983.hg.1 |
ENST00000541806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006983.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006983.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006983.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006982.hg.1 |
TC0X000521.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006982.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006982.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006982.hg.1 |
ENST00000541806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006982.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006982.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006982.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006978.hg.1 |
TC0X000521.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006978.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006980.hg.1 |
TC0X000521.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006980.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006980.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006981.hg.1 |
TC0X000521.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006981.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006981.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006981.hg.1 |
ENST00000541806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006981.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006981.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006981.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008056.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155114.hg.1 |
TC0X00008056.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155112.hg.1 |
TC0X00008056.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155111.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155111.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155111.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155111.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155111.hg.1 |
TC0X00008056.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
ENST00000336803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
ENST00000540876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
ENST00000541806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
NM_001171092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
NM_001171095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155113.hg.1 |
TC0X00008056.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155109.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155109.hg.1 |
NM_020384.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155109.hg.1 |
TC0X00008056.hg.1 |
Clariom D TC |