Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000597441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000597441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000324041.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000431178.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000593885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000596876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000597441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000597441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000598305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000599865.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
ENST00000602148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_001302961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NM_004917.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011226.hg.1 |
NR_126566.1 |
Refseq |