Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306129.1 | NP_001293058.1 | 44 - 45 | 7324 | 72 | |
NM_001306130.1 | NP_001293059.1 | 42 - 43 | 6787 | 72 | ||
NM_001306131.1 | NP_001293060.1 | 42 - 43 | 6781 | 72 | ||
NM_001306132.1 | NP_001293061.1 | 42 - 43 | 6706 | 72 | ||
NM_002026.3 | NP_002017.1 | 43 - 44 | 7049 | 72 | ||
NM_212474.2 | NP_997639.1 | 41 - 42 | 6512 | 72 | ||
NM_212476.2 | NP_997641.1 | 41 - 42 | 6872 | 72 | ||
NM_212478.2 | NP_997643.1 | 43 - 44 | 6974 | 72 | ||
NM_212482.2 | NP_997647.1 | 43 - 44 | 7415 | 72 | ||
XM_005246397.1 | XP_005246454.1 | 43 - 44 | 7514 | 72 | ||
XM_005246398.1 | XP_005246455.1 | 43 - 44 | 7439 | 72 | ||
XM_005246399.1 | XP_005246456.1 | 44 - 45 | 7421 | 72 | ||
XM_005246401.1 | XP_005246458.1 | 44 - 45 | 7346 | 72 | ||
XM_005246402.1 | XP_005246459.1 | 44 - 45 | 7346 | 72 | ||
XM_005246403.1 | XP_005246460.1 | 42 - 43 | 7244 | 72 | ||
XM_005246404.1 | XP_005246461.1 | 42 - 43 | 7241 | 72 | ||
XM_005246405.1 | XP_005246462.1 | 42 - 43 | 7244 | 72 | ||
XM_005246406.1 | XP_005246463.1 | 42 - 43 | 7241 | 72 | ||
XM_005246407.1 | XP_005246464.1 | 43 - 44 | 7154 | 72 | ||
XM_005246408.1 | XP_005246465.1 | 43 - 44 | 7151 | 72 | ||
XM_005246409.1 | XP_005246466.1 | 42 - 43 | 7166 | 72 | ||
XM_005246410.1 | XP_005246467.1 | 43 - 44 | 7076 | 72 | ||
XM_005246411.1 | XP_005246468.1 | 42 - 43 | 6884 | 72 | ||
XM_005246412.1 | XP_005246469.1 | 41 - 42 | 6896 | 72 | ||
XM_005246414.1 | XP_005246471.1 | 42 - 43 | 6881 | 72 | ||
XM_005246416.1 | XP_005246473.1 | 42 - 43 | 6803 | 72 | ||
XM_017003692.1 | XP_016859181.1 | 43 - 44 | 7154 | 72 | ||
XM_017003693.1 | XP_016859182.1 | 43 - 44 | 7151 | 72 | ||
XM_017003694.1 | XP_016859183.1 | 43 - 44 | 7076 | 72 | ||
XM_017003695.1 | XP_016859184.1 | 41 - 42 | 6971 | 72 | ||
GenBank mRNA | AB191261.1 | - | 42 - 43 | 72 | ||
AB209287.1 | - | 19 - 20 | 72 | |||
AB209840.1 | - | 42 - 43 | 6771 | 72 | ||
AF130095.1 | - | 7 - 8 | 1078 | 72 | ||
AJ320525.1 | - | 8 - 9 | 72 | |||
AJ320526.1 | - | 7 - 8 | 72 | |||
AJ320527.1 | - | 8 - 9 | 72 | |||
AK297733.1 | - | 9 - 10 | 1196 | 72 | ||
AK300216.1 | - | 4 - 5 | 775 | 72 | ||
AK300246.1 | - | 18 - 19 | 72 | |||
AK300458.1 | - | 21 - 22 | 72 | |||
AK316350.1 | - | 6 - 7 | 833 | 72 | ||
AL832202.1 | - | 41 - 42 | 6513 | 72 | ||
AL832771.1 | - | 41 - 42 | 6513 | 72 | ||
BC005858.2 | - | 3 - 4 | 352 | 72 | ||
BC016875.2 | - | 4 - 5 | 72 | |||
BC117176.1 | - | 41 - 42 | 6492 | 40125727 | 72 | |
BC143754.1 | - | 42 - 43 | 6684 | 9052268 | 72 | |
BC143763.1 | - | 41 - 42 | 6492 | 9052277 | 72 | |
BT006856.1 | - | 2 - 3 | 207 | 72 | ||
BX537590.1 | - | 44 - 45 | 7324 | 72 | ||
BX538017.1 | - | 43 - 44 | 7050 | 72 | ||
BX538018.1 | - | 43 - 44 | 7050 | 72 | ||
BX640608.1 | - | 41 - 42 | 6872 | 72 | ||
BX640638.1 | - | 19 - 20 | 72 | |||
BX640731.1 | - | 43 - 44 | 7050 | 72 | ||
BX640802.1 | - | 43 - 44 | 6973 | 72 | ||
BX640803.1 | - | 43 - 44 | 7416 | 72 | ||
BX640875.1 | - | 43 - 44 | 72 | |||
BX640920.1 | - | 23 - 24 | 3485 | 72 | ||
BX640999.1 | - | 39 - 40 | 6298 | 72 | ||
BX641150.1 | - | 17 - 18 | 8151 | 72 | ||
BX649182.1 | - | 43 - 44 | 6973 | 72 | ||
CR749281.1 | - | 42 - 43 | 6818 | 72 | ||
CR749316.1 | - | 42 - 43 | 6784 | 72 | ||
CR749317.1 | - | 42 - 43 | 72 | |||
CR749666.1 | - | 23 - 24 | 3664 | 72 | ||
CR936623.1 | - | 20 - 21 | 72 | |||
M10905.1 | - | 12 - 13 | 72 | |||
U42456.1 | - | 2 - 3 | 72 | |||
U42592.1 | - | 2 - 3 | 72 | |||
X02761.1 | - | 43 - 44 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |